Learn More
Teledyne ISCO, Inc Cp Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.00 - 24.00 Ml/Min.; 10,000 Psi
Supplier:  Teledyne ISCO, Inc CP024SRX
Encompass
Request Bulk Order
CP Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 24.00 mL/min.; 10,000 psi; Stainless Steel Fluid Path; without Piston Wash; with Pressure & Leak Sensors (1,000 psi max), no dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9935
Explore available promotions