Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1 Mouse, RD1, Isotype control, Beckman Coulter™
Supplier: Beckman Coulter 6602884
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Description
Host Species: Mouse
Clone: 2T8-2F5
Isotype: IgG1
Species Reactivity: Human
Specifications
2T8-2F5 | |
Mouse | |
Human | |
IgG1 |
RD1 | |
100 Tests | |
Purified |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction