Learn More
Ted Pella Inc COPPER TAPE COND.SGL ADH.
Supplier: Ted Pella Inc 16072
Copper Conductive Tape, 6.3mm W x 16.4m L; 1/4 in. x 18yd, Core diameter is 76mm, Foil Thickness: 0.04mm, Total Thickness: 0.07mm, Resistance through Adhesive: 0.005ohm