Learn More
Ted Pella Inc COPPER CONDUCTIVE TAPE 12.7MM
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ted Pella Inc 160721
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tape, Conductive; Copper; 3M; Single Adhesive Surface; 12.7mm; The tape offers excellent conductivity through the foil backing; Foil Thickness: 0.04mm; Total Thickness: 0.07mm; Resistance through Adhesive: 0.005 ohm