Learn More
Mopec™ Control Slides, Special Stains, Helico Bacteria (Pylori HP)
All positive control slides are on Mopec™ IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
$158.40 - $840.96
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
22-457-742
|
Mopec™
SC81410 |
10 Unstained Slides, 1 Stained Slide |
Case of 10 for $158.40
|
|
|||||
22-457-433
|
Mopec™
SC81420 |
20 Unstained Slides, 2 Stained Slides |
Case of 20 for $273.60
|
|
|||||
22-457-621
|
Mopec™
SC814100 |
98 Unstained Slides, 2 Stained Slides |
Case of 100 for $840.96
|
|
|||||
Description
- Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
- Case of 10 contains 10 unstained slides and 1 stained slide
- Case of 20 contains 20 unstained slides and 2 stained slides
- Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Control Slide |