Learn More
Perkin Elmer US LLC Continuous Wave Module with Upper Bracket and Mirror - 150 W
Supplier: Perkin Elmer US LLC N4202036
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our Fluorescence Spectrometers use Pulsed Xenon Discharge lamps. These reliable, stable lamps provide remarkable sensitivity and can achieve a peak intensity exceeding 1 KW. For optimal instrument perfomance, always use original PerkinElmer replacement lamps.