Learn More
Quadrex Corporation COLUMN NONPOLAR 007-1 DIMETHYL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Quadrex Corporation 007110V5.0F
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
non-polar 007-1 dimethylpolysiloxane phase column, 10 meter, 0.53 mm internal diameter, film thickness 5.00 microns
This item is not returnable.
View return policy