Learn More
Teledyne ISCO, Inc RediSep Gold Silica Gel Disposable Flash Columns, 80 gram. Package of 6.
Supplier: Teledyne ISCO, Inc 692203348
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
High performance flash column which delivers superior sample purity through the use of fine spherical silica gel (20-40 µm particle size). Equipped with RFID technology, allowing Teledyne ISCO CombiFlash systems with RFID to automatically detect the column type and size.
This item is not returnable.
View return policy