Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Reagent for Nitrogen (Nitrate), For Automated Cadmium Reduction Method, LabChem™
$110.00 - $468.00
Specifications
Color | Clear, slight pink to tan |
---|---|
Format | Liquid |
Packaging | Amber Glass |
Storage Requirements | Refrigerate |
Type | Color Reagent for Nitrogen (Nitrate) |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
LC132801
|
LabChem, Inc.
LC132801 |
500 mL | Amber Glass |
Each for $110.00
|
|
||||
LC132802
|
LabChem, Inc.
LC132802 |
1 L | Amber Glass |
Each for $168.00
|
|
||||
LC132804
|
LabChem, Inc.
LC132804 |
4 L | Amber Glass |
Each for $468.00
|
|
||||
Specifications
Clear, slight pink to tan | |
Amber Glass | |
Color Reagent for Nitrogen (Nitrate) |
Liquid | |
Refrigerate |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title