Learn More
Cole-Parmer Cole-Parmer PP Wall Mount Drying Rack with Pegs, 20 Place
Supplier: Cole-Parmer 0604591
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Polypropylene draining rack provides an excellent option for post-wash draining of glassware. Labware dries quickly on screw-in pegs. Rack features 5 tiers of 20 pegs, hooks for wall mounting, a wastewater tray, and draining port.