Learn More
Grainger Coin Cell Battery, 2025 Battery
Supplier: Grainger 2CUR8
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Panasonic CR2025 lithium coin cell battery has a 3V nominal voltage and 225 mAh capacity. It has a stainless steel can construction and measures 20mm in diameter by 2.5mm thick. The battery has a 10 year shelf life and operating temperature range of -30°C to +70°C. It features low self-discharge and high drain/pulse capability, making it well-suited for devices like watches, calculators, car key fobs, and other electronics needing button cell batteries. The Panasonic CR2025 offers reliable, long-lasting power in a compact coin cell form factor.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.