Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M Coban™ Self-Adherent Wrap
Coban™ Self-Adherent Wrap - Conformable, lightweight, and cohesive
Supplier: 3M MMM 1581
This item is not returnable.
View return policy
Description
- Self-adherent wrap may be used for compression dressings and for securing dressings and other devices
- Functions like a tape without adhesives; no pins or clips are needed
- Packaged in 5 yd. (4.6 m) rolls
Specifications
Elastic | |
1 in. x 5 yd. | |
Tan | |
No |
Self-Adherent Wrap | |
Overwrap for Splints, Support Sprains and Strains, Secure Dressings and Devices, Compression to Reduce Edema, Reduce Post-Operative Swelling | |
Self Adhesive |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction