Learn More
Electron Microscopy Sciences Coated Lead Shield
Supplier: Electron Microscopy Sciences 64460
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This 1/4" (6.4 mm) thick lead shield protects against chemical reactions and nuclear radiation. The shield is coated with vinyl for a soft smooth surface, which eliminates the hazard of any skin contact as well as flying debris. S