Learn More
STEMCELL Technologies CMV (pp65) Peptide Pool, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 1000668
Encompass_Preferred
The CMV (pp65) Peptide Pool is a lyophilized mixture of 138 peptides from the 65 kDa phosphoprotein (pp65) of human cytomegalovirus (CMV; strain AD169). pp65 has been shown to counteract the induction of interferon response by preventing the activation of interferon response factor 3 (IRF-3). pp65 is also important in inducing transcription of viral immediate-early promoter. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 561 on pp65.
Catalog No. NC1962505
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov