Learn More
Cleatech SDPVC 24" FLOW HOOD W/ STND
Supplier: Cleatech 10007HFLS
Portable Bench top Vertical Laminar Flow station, Work space 24.5"Wx 18.5"Dx 19.75"H ,Static- dissipative PVC construction, Static- dissipative PVC front tilt-up sash, 110VAC, 50/60Hz. Adjustable speed Impeller, Backward Curve, 730 CFM @ 0.000-In,Including 30" Stand , Including HEPA Filter , Including Built-in Fluorescent Light