Learn More
Electron Microscopy Sciences Circlips Insertion Tool
Supplier: Electron Microscopy Sciences EMS016
Circlips are for all EMS TEM sample holders and specimen rods. They are made from heat treated Beryllium copper. The Circlip extraction tool is for removing our Circlips from all TEM sample holders.