Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Berkshire™ Choice™ Cleanroom Face Masks
Three-layer cleanroom face masks provide filtration efficiency while providing long wearing comfort.
$1345.00 - $2880.00
Specifications
Product Type | Cleanroom Face Mask |
---|---|
Color | White |
Fastening Type | Earloops |
Material | Polypropylene |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Packaging Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Type | Price | Quantity & Availability | |||||
19-266-708
|
Berkshire™
CHFM18040B |
Bulk Packaged |
Case of 2000 for $2,880.00
|
|
|||||
CHFM18024I
|
Berkshire™
CHFM18024I |
Individually Bagged |
Case of 600 for $1,345.00
|
|
|||||
Description
- 3-Layer cleanroom masks
- Knitted nylon earloops
- Filter medium: meltblown polypropylene provides particle and bacterial filtration
- Fully enclosed nosepiece is adjustable
- Available in bulk or individually wrapped packaging
Specifications
Cleanroom Face Mask | |
Earloops | |
Non-sterile | |
ASTM F2299 | |
Class 10 (ISO 4) | |
Single-use | |
Choice™ |
White | |
Polypropylene | |
Pleated | |
EN 14683 | |
3-ply | |
99.7% (PFE), 99.7% (BFE) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title