Learn More
Enterprise Technology Solutions Chemical Safety Goggles, Clear Lens
Supplier: Enterprise Technology Solutions CRW2230RBX
Provides a wide, unobstructed field of vision.Can be worn with most half-mask respirators.Transparent protective PVC body.Comfortable fit over prescription eyewear.Protects against impact and splash.