Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHEMetrics COD Calibration Standard, 10,000 ppm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For use with COD (Chemical Oxygen Demand) in measuring the overall level of organic contamination in wastewater
Supplier: CHEMetrics A7310
Description
- COD Calibration Standard
- 10,000 ppm
- 200 mL
- Shelf-Life: 8 months
Specifications
Recycling Container | |
For disposal of CHEMetrics mercury-containing COD vials |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Not available in all areas.
Spot an opportunity for improvement?
Provide Content Correction