Learn More
Chembio DPP™ HIV 1/2 Rapid Test Control Pack
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Control Pack for Chembio DPP™ HIV 1/2 Assay
Supplier: Chembio 6095520
This item is not returnable.
View return policy
Description
Chembio DPP™ HIV 1/2 Assay Features:- Patented DPP™ technology for the rapid detection of HIV 1 and HIV 2 antibodies in oral fluid and all blood matrices
- Intended for use as a point-of-care test to aid in the diagnosis of infection with HIV 1 and HIV 2
- Sharp and distinctive visual results
- Excellent sensitivity and specificity
- Built-in procedural control to human IgG
- SampleTainer™ bottle: a safe, closed system for handling potentially infectious blood and oral fluid samples
Specifications
1 pack contains:1 HIV-1 Reactive Control 0.5 mL;1 HIV-2 Reactive Control 0.5 mL;1 HIV Nonreactive Control 0.5 mL; Each kit control vial contains sufficient volume to run 50 tests |