Learn More
Coastline Global™ CheckUp Pro Dog - At home wellness test for dogs
CheckUp Pro dog urine test strips provide a quick, simple, and reliable testing method to observe the general wellness of dogs through at-home urine testing. The CheckUp pro also make urine sample collection stress-free and easy.
Supplier: Coastline Global™ CUDPRO
Description
At home urine test kit for dogs comes with necessary collection supplies to monitor your dog's wellness.
- Telescopic pole and Disposable cup
- 2 identical testing strips -- repeat test with second strip if necessary
- Sample collection pipette
- Sample collection vial
Specifications
Self-declared | |
Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes | |
Ready-to-Use (RTU) |
Waived (Urine) | |
Single-use |