Learn More
Coastline Global™ CheckUp Pro Cat - At home wellness test for cats
CheckUp Pro cat urine test strips provide a quick, simple and reliable testing method to observe the general wellness of cats through at-home urine testing. The CheckUp pro also make urine sample collection stress-free and easy.
Supplier: Coastline Global™ CUCPRO
Description
At-home urine testing for general cat wellness made easy with this quick, reliable, and simple testing kit.
- 2 lbs (900 g) hydrophobic litter
- 2 identical testing strips -- repeat test with second strip if necessary
- Sample collection pipette
- Sample collection vial
Specifications
Self-declared | |
Glucose, Bilirubin, Ketone, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes | |
Ready-to-Use (RTU) |
Waived (Urine) | |
Single-use |