Learn More
Ace Glass, Inc. CHECK VALVE 8MMCHECK VALVE 8 CHECK VALVE 8MMCHECK VALVE 8
Supplier: Ace Glass, Inc. 877409
All-glass check valve with ground seat and precision ground solid ball. Suitable with mercury in the vertical position, more sensitive to other fluids in the horizontal position. Connections are 8mm O.D.