Learn More
Premier Lab Supply CEMBLES Pre-Assembled XRF Sample Cups – PN# PA-120-32-135
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Premier Lab Supply PA12032135
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
CEMBLES Pre-Assembled XRF Sample Cups! (CEMBLES PN# PA-120-32-135; 32mm Cups PN# SC-3332; Mylar film 3.5μm; 48 per pack). CEMBLES are custom made to your specifications & available in variant configurations of cups and thin films that have been pre-assembled for your convenience.