Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Skid-Resistant Spunbond Polypropylene Shoe Covers

Catalog No. M28033
Click to view available options
Size:
Universal
X-Large
Packaging Type:
Bags
Dispenser Boxes
This item is not returnable. View return policy
This item is not returnable. View return policy

Skid-resistant shoe covers provide lightweight protection from non-hazardous liquids and dry particulates. Ideal for visitors or light duty applications.

  • Sponbond polypropylene
  • Non-conductive
  • Lightweight
  • 150 pair per case, bags or boxes

Specifications

Size X-Large
Material Spunbond Polypropylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Sole Type Non-skid
Model 28033
For Use With (Application) Visitors, Light Duty, General Purpose Maintenance, Cleaning
Packaging Quantity 150 Pairs/Cs.
Packaging Type Dispenser Boxes
Resistance Fluid
Product Type Shoe Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.