Learn More
Bioassay Systems CellQuanti-Blue™ Cell Viability Assay Kit (10000 tests). Homogeneous assay for cell viability, proliferation, cytotoxcity and high-throughput screen for anticancer agents.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems CQBL10K
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Homogeneous assay for cell viability, proliferation, cytotoxcity and high-throughput screen for anticancer agents. Key Features: Safe. Non-radioactive assay (cf. 3H-thymidine incorporation assay). Sensitive and accurate. As low as 100 cells can be accurately quantified. Time efficient. High-throughput assay in 96-well and 384-well plates allows simultaneous processing ten of thousands of samples per day. Homogeneous and convenient. A single reagent and "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS. Z factors of 0.6 to 0.9 are routinely observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems. Method: FL530/590nm. Samples: Cell culture. Species: All. Procedure: Assay takes 1-5 hrs. Kit size: 10000 tests. Detection limit: 100 cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.