Learn More
Cell Biolabs Inc CELL TRANSFRM ASSAY 96 WELL
Supplier: Cell Biolabs Inc CBA130
The CytoSelect 96-Well Cell Transformation Assay (Soft Agar Colony Formation) quantifies colonies with no manual counting required. Cells are incubated in soft agar for 7-8 days and then quantified using the included fluorescent dye.