Learn More
Biotang Inc Disposable Delta* Cell Spreader, 1/PK, 500/CS, sterile, Uniquely shaped spreading bar has an innovative edgeless design. The delta-shaped head distributes pressure evenly for smooth spreading. Individually bagged. Measures 172 x 37 x 14 mm.
Supplier:  Biotang Inc HD8151
Encompass_Preferred
Disposable Delta* Cell Spreader, 1/PK, 500/CS, sterile, Uniquely shaped spreading bar has an innovative edgeless design. The delta-shaped head distributes pressure evenly for smooth spreading. Individually bagged. Measures 172 x 37 x 14 mm.
Catalog No. 50-751-5036
Explore available promotions