Learn More
Alkali Scientific Cell Scrapers, Blade Width 20mm, Length 290mm, Sterile, 100/cs
SDP
Supplier:  Alkali Scientific C30020
Encompass_Preferred
Alkali Scientific Cell Scrapers feature a rotatable blade for mechanical harvesting of adherent cells from tissue culture containers. They are individually packaged, gamma sterilized, and pyrogen free.
Catalog No. 50-233-5948
Explore available promotions