Learn More
Sarstedt Inc Cell scraper, 2-position blade, size: S
Supplier:  Sarstedt Inc 83.3950
Encompass_Preferred
Cell scraper, 2-position blade, size: S, handle length: 24 cm, blade length: 1.35 cm, sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, 1 piece(s)/blister.
Catalog No. NC1890482
Explore available promotions