Learn More
Sarstedt Inc Cell scraper, 2-position blade, size: M
Supplier: Sarstedt Inc 83.3951
Cell scraper, 2-position blade, size: M, handle length: 24 cm, blade length: 1.7 cm, sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, 1 piece(s)/blister.
This item is not returnable.
View return policy