Learn More
CELLTREAT Scientific Products Cell Scraper, 25cm Handle, 20mm Scraper Blade, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229310
Encompass_Preferred
CELLTREAT Scientific Products 25cm Cell Scraper. Cell Scrapers are designed to gently harvest cells from tissue culture flasks, dishes, or bottles. Soft, pliable blade won't damage cells or scratch surfaces. Scraper blade is positioned parallel to the handle for windshield wiper like scraping. Handle length 25cm and blade length 20mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped, 100/case.
Catalog No. 50-201-974
Explore available promotions