Learn More
Bioworld Cell Lifter, Sterile, 18cm Handle, 1 CS
Supplier: Bioworld 309000181
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These lifters provide a convenient method for removal of cells from a variety of cell culture containers.Handle Length: 18cm Blade Length: 2.0cm Polyethylene Disposable Gamma Radiation SterilizedNon-PyrogenicIndividually Wrapped 100/case