Learn More
Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassays, Calibrators and Controls
Complete your ISD panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay.
$529.00 - $540.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Detectable Analytes | Mycophenolic Acid |
Sample Type | Plasma |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
23-278-295
|
Thermo Scientific™
100276 |
CEDIA Mycophenolic Acid Assay |
Each for $4,434.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-296
|
Thermo Scientific™
10016265 |
CEDIA Mycophenolic Acid Assay for Indiko |
Each for $4,995.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-290
|
Thermo Scientific™
100277 |
CEDIA Mycophenolic Acid Calibrator Set |
Each for $540.00
|
|
|||||
23-278-437
|
Thermo Scientific™
100278 |
CEDIA Mycophenolic Acid Control 1 |
Each for $529.00
|
|
|||||
23-278-439
|
Thermo Scientific™
100280 |
CEDIA Mycophenolic Acid Control 3 |
Each for $538.00
|
|
|||||
Description
Complete your immunosuppressant drug testing panel with the Thermo Scientific™ CEDIA™ Mycophenolic Acid Immunoassay. Like our other ISD assays, the CEDIA Mycophenolic Acid Immunoassay is accurate and precise, providing users with confidence in their results. Applications are available on our flexible and intuitive Indiko™ and Indiko Plus clinical and specialty chemical systems as well as a wide variety of other laboratory instrumentation.
Key Features- Optimal low-end sensitivity with a broad dynamic range
- Excellent correlation with HPLC-MS, the gold standard method
- Precise throughout the standard curve assay range
Specifications
2°C to 8°C | |
Mycophenolic Acid | |
Plasma |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.