Learn More
Thermo Scientific™ CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays.
$671.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Multi-Drug Controls |
Detectable Analytes | Amphetamine/MDMA |
DoA Calibrators | CEDIA Multi-drug Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-202
|
Thermo Scientific™
100103 |
1 x 65 mL, MCC |
Each for $1,736.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-200
|
Thermo Scientific™
10016417 |
3 x 17 mL, Indiko |
Each for $671.00
|
|
|||||
23-278-201
|
Thermo Scientific™
100104 |
3 x 17 mL, LC |
Each for $1,640.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of Thermo Scientific™ CEDIA™ Amphetamine/Ecstasy Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA™ assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs.
- One of the largest menus of tests available
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Amphetamine/MDMA | |
Urine |
MGC Multi-Drug Controls | |
CEDIA Multi-drug Calibrators | |
Reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.