Learn More
Vileda Professional™ CE MicroIntensive Duo™
Double-sided, single-use mop to speed-up the cleaning process, while also reducing the risk of cross-contamination.
$792.50 - $1001.00
Specifications
Color White
Autoclavable Autoclavable
Material Polyester Microfiber
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
19-200-827
View Documents
Vileda Professional™
166813
Non-sterile
Case of 25 for $792.50
Add to Cart
19-200-828
View Documents
Vileda Professional™
166815
Sterile
Case of 25 for $1,001.00
Add to Cart
Description
Description
  • Independent lab study, conducted by BMA Labs, resulting in bacteria reduction of 99.99%
  • 4-layer construction - featuring Polyester/Microfiber embossed cleaning side
  • Pre-laundered for single-use
  • Gamma and autoclave compatible
  • Extremely low particle shedding
  • Double-sided cleaning with color -stitched side indicator
  • Corner drainage openings for cleaner rinses
  • Finger loop for touchless on/off of cleaning sides
Specifications
Specifications
White
Autoclavable
Polyester Microfiber