Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD6 Mouse anti-Rat, BV510, Clone: OX-52, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 745045
Description
CD6 Monoclonal antibody specifically detects CD6 in Rat samples. It is validated for Flow Cytometry.Specifications
CD6 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Rat | |
Purified |
Flow Cytometry | |
OX-52 | |
Brilliant Violet 510 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction