Learn More
Ted Pella Inc Carbon Tape, 8mm W x 20m L
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ted Pella Inc 16073
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Both sides of the tape have a thick conductive adhesive (conductive inner film is 35µm and the adhesive is 45µm on each side for a total of 125µm [5 mils]) with a white liner. The double adhesive and conductive design permits quick mounting of samples without using liquid or colloidal adhesives. The tape is relatively solid and non-porous and does not absorb small samples. The variety of widths affords efficient application to various specimen mount surface sizes. Thickness is 0.16mm. Available in six widths. Can be used down to -20°C (-4°F) and temperature maximum is 60°C (140°F). Available in 5, 8, 12, 20, 25, and 50mm W x 20m L (0.19, 0.31, 0.47, 0.79, 0.98, and 1.96" x 21.8 yd). Core diameter is 76mm (3"). SEM mounts not included.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.