Learn More
Restek™ Capillary Ferrules
SDP
For 1/16 in. Compression-Type Fittings, High Purity & High Density
$51.60 - $319.50
Specifications
Type Ferrule
View More Specs
Products 18
Catalog Number Mfr. No. Diameter (Metric) Material For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Material For Use With (Equipment) Quantity Price Quantity & Availability  
06-717-639
SDP
View Documents
Restek™
20231
0.5 mm Graphite, Vespel Fits Column I.D. 0.32 mm 50 Pk.
Pack of 50 for $319.50
Add to Cart
06-717-621
SDP
View Documents
Restek™
20211
0.04 cm Graphite, Vespel 0.062 in. Compression Type Fitting, Fits Column 0.1, 0.15, 0.18, 0.25, 0.28 mm 10 Pk.
Pack of 10 for $87.20
Add to Cart
06-717-623
SDP
View Documents
Restek™
20213
0.08 cm Graphite, Vespel 0.062 in. Compression Type Fitting, Fits Column I.D. 0.45, 0.53 mm 10 Pk.
Pack of 10 for $88.20
Add to Cart
06-717-622
SDP
View Documents
Restek™
20212
0.05 cm Graphite, Vespel 0.062 in. Compression Type Fitting, Fits Column I.D. 0.28, 0.32 mm 10 Pk.
Pack of 10 for $88.20
Add to Cart
06-611-126
SDP
View Documents
Restek™
21058
1 mm Graphite Capillary Columns 10 Pk.
Pack of 10 for $55.00
Add to Cart
06-717-637
SDP
View Documents
Restek™
20229
0.04 cm Graphite, Vespel 0.062 in. Compression Type Fitting, Fits Column 0.1, 0.15, 0.18, 0.25, 0.28 mm 50 Pk.
Pack of 50 for $319.50
Add to Cart
06-611-127
SDP
missing translation for 'viewDocuments'
Restek™
21060
1.6 mm Graphite Capillary Columns 10 Pk.
Pack of 10 for $51.60
Add to Cart
06-611-143
SDP
missing translation for 'viewDocuments'
Restek™
22214
0.4 mm Vespel 0.10/0.15/0.18/0.25/0.28 mm ID Columns 10 Pk.
Pack of 10 for $73.90
Add to Cart
06-611-144
SDP
missing translation for 'viewDocuments'
Restek™
22216
0.8 mm Vespel 0.45/0.53 mm ID Columns 10 Pk.
Pack of 10 for $73.90
Add to Cart
06-611-145
SDP
missing translation for 'viewDocuments'
Restek™
22217
1 mm Vespel 0.75 mm ID Columns 10 Pk.
Pack of 10 for $71.30
Add to Cart
06-611-100
SDP
missing translation for 'viewDocuments'
Restek™
20275
0.3 mm 40% Graphite, 60% Vespel Capillary Columns 10 Pk.
Pack of 10 for $68.00
Add to Cart
06-611-098
SDP
missing translation for 'viewDocuments'
Restek™
20232
0.6 mm 40% Graphite, 60% Vespel 0.28 mm ID Columns 10 Pk.
Pack of 10 for $70.80
Add to Cart
06-611-103
SDP
missing translation for 'viewDocuments'
Restek™
24912
1 mm 40% Graphite, 60% Vespel Micropacked Columns 10 Pk.
Pack of 10 for $71.60
Add to Cart
06-611-104
SDP
missing translation for 'viewDocuments'
Restek™
27061
0.3 mm 15% Graphite, 85% Vespel Capillary Columns 10 Pk.
Pack of 10 for $68.00
Add to Cart
06-611-105
SDP
missing translation for 'viewDocuments'
Restek™
27062
0.4 mm 15% Graphite, 85% Vespel Capillary Columns 10 Pk.
Pack of 10 for $74.20
Add to Cart
06-611-106
SDP
missing translation for 'viewDocuments'
Restek™
27066
0.6 mm 15% Graphite, 85% Vespel 0.25 mm MXT™ Columns 10 Pk.
Pack of 10 for $73.70
Add to Cart
06-611-107
SDP
missing translation for 'viewDocuments'
Restek™
27067
0.8 mm 15% Graphite, 85% Vespel Capillary Columns 10 Pk.
Pack of 10 for $82.00
Add to Cart
06-611-109
SDP
missing translation for 'viewDocuments'
Restek™
27069
1.2 mm 15% Graphite, 85% Vespel Micropacked Columns 10 Pk.
Pack of 10 for $69.30
Add to Cart
Description
Description
Vespel Ferrules
  • 100% High-Temperature Polyimide
  • Durable, leak tight
  • Stable to 350°C
Graphite Ferrules
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Smoother surface and cleaner edges than conventional graphite ferrules
  • Contain no binders that can off-gas or adsorb analytes
  • Stable to 450°C
Vespel/Graphite Ferrules
  • VG2 offers the best combination of sealing and ease of workability
  • VG1 offers equivalent composition to Agilent Vespel/graphite ferrules
  • Seal with minimal torque
  • Preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
  • Reusable
  • Stable to 400°C
Specifications
Specifications
Ferrule