Learn More
Cole-Parmer Fluke 725 US Multifunction Process Calibrator
Supplier:  Cole-Parmer 2607250
Encompass_Preferred
Measure and source functions o fthis process calibrator allow you to test and calibrate almost anything! It has easy-to-read measure and source screen to allow view input and output at the same time. The calibrator can perform rapid linearity tests with auto step and auto ramp features. Its rugged, streamlined design is ideal for field conditions.
Catalog No. 50-993-328