Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Industrial Scientific Calibration Gas
Keeps gas monitoring instruments operating accurately and reliably with complete gas calibration kits and replacement cylinders
$180.00 - $1220.00
Catalog Number | Mfr. No. | Concentration | Components Measured | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Components Measured | Capacity (Metric) | Price | Quantity & Availability | |||
19-999-231
|
Industrial Scientific
18100701 |
100ppm | Carbon Monoxide | 34 L |
Each for $180.00
|
|
|||
19-999-282
|
Industrial Scientific
18101477 |
25ppm | Nitrogen Dioxide | 58 L |
Each for $1,220.00
|
|
|||
18-999-4503
|
Industrial Scientific
18102320 |
Oxygen 20.9%, carbon monoxide 100ppm | Oxygen, Carbon Monoxide | 552 L |
Each for $848.50
|
|
|||
18-301
|
Industrial Scientific
18100859 |
25ppm | Hydrogen Sulfide | 58 L |
Each for $723.00
|
N/A | |||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Catalog Number 18-298 is a kit containing the gas cylinder as well as a 0.5lpm Regulator (Mfr. No. 18100883)
Specifications
Calibration Gas Cylinder |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title