Learn More
Abcam Calcium Assay Kit (Colorimetric) (ab102505) 250 tests
Supplier:  Abcam AB102505
Encompass
Assay type: QuantitativePlatform: Microplate readerAssay time: 20 minSample type: Cell Lysate, Other biological fluids, Plasma, Serum, Tissue Extracts, Urine
Catalog No. NC2958236