Learn More
Wilmad Labglass BURET AUTO LAB GRADE 25ML
Supplier: Wilmad Labglass LG4241T102
Buret, Laboratory Grade, Automatic, with Straight PTFE Stopcock, Complete. Buret Capacity: 25mL; Subdivision: 0.1mL; Reservoir Capacity: 16 ounces; Stopcock Size: 2mm. Buret is filled by pressure supplied by the rubber bulb.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov