Learn More
Bullard™ Sampling Connector Kit for COHP CO Monitor
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Kit connects the COHP monitor to the air source.
Supplier: Bullard™ COHPKIT
This item is not returnable.
View return policy
Description
- Sampling connector kit includes:
- 3/8 in. x 1/4 in. adaptor
- Colder coupler tee fitting
- Reinforced sampling tube with colder plugs – 5 ft. length
Specifications
3/8 in. x 1/4 in. adapter, colder coupler tee fitting, reinforced sampling tube with colder plugs – 5 ft. length | |
COHPKIT |