Learn More
Bullard™ Replacement Inlet Filter Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Medium efficiency replacement filter for use with fresh air pumps.
Supplier: Bullard™ 23612
This item is not returnable.
View return policy
Description
- Replacement inlet filter
- Keeps moisture and particulates out of air supply
- Helps protect air pumps from carbon vanes
- Regular inspection and replacement of air filters will prevent extensive and costly supplied air pump repair
- Dirty or clogged filters can be responsible for failure of the pump to build up outlet pressure and eventual overheating
- Recommended replacement is every 500 running hours or sooner if necessary
Specifications
23612 |