Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Replacement Filters for 100 CFM Clean Air Box Series Breathing Air Filtration Systems
SDP

Filter element replacement parts for use with 100 CFM Clean Air Boxes.

Supplier:  Bullard™ CAB100FEB

 View more versions of this product

Catalog No. 19-100-689


Add to Cart

Description

Description

  • Replacement filter elements for Clean Air Boxes (CAB)
  • For use with 100 cfm models only
  • Choose stage 1, 2, or 3 separate element replacement parts or element kit including all stages 1, 2, and 3
Specifications

Specifications

100 cfm
Element B-Coalescing
Replacement Filter
Clean Air Box Series 100 cfm
CAB100FEB
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit