Learn More
Bullard™ Wall Unit Clean Air Box (CAB) Series Fixed Breathing Air Filtration/CO Monitoring, One or Two Worker System
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Panel mount manifold provides 3-stage air filtration and CO monitoring to help satisy OSHA Grade D breathing air requirements.
Supplier: Bullard™ CAB30PMIRSS
Description
- Durable construction withstands harsh working environments
- Powder coated steel panel mount unit
- Independent Regulators
- Auto condensate drains will extend the filter element lifespan
- CO Monitor
Specifications
CAB30PMIRSS |