Learn More
Pfanstiehl Inc Sodium Succinate, Hexahydrate, High Purity Low Endotoxin, NF, JPE (D-161) – 1kg pack – GMP, Multicompendial – CAS: 6106-21-4
Supplier: Pfanstiehl Inc D1611KG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sodium Succinate (Hexahydrate) is the fully hydrated form of sodium succinate. Sodium Succinate is generally used as a Biopharmaceutical ingredient for pH buffering and final product formulation (e.g. Antibody Drug Conjugates/ADCs). The physical properties of this product make it a suitable tool for use in a formulation. Pfanstiehl High Purity / Low Endotoxin-Low Metal Sodium Succinate (Hexahydrate) was developed specifically for biopharmaceutical formulation and commercial manufacturing and produced under full cGMP conditions in the United States to the highest quality and purity standards. Pfanstiehl Brand Sodium Succinate (Anhydrous) is compliant with the USP monograph established in 2017 and available as USP-NF (United States Pharmacopoeia—National Formulary).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.