Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Danger: Poison with Pictogram Signs
For identifying equipment, rooms and materials that contain or are contaminated with hazardous agents
$13.50 - $66.00
Specifications
Quantity | 1 Ea. |
---|---|
Min. Temperature (English) | -40°F |
Displayed Text | DANGER Poison |
Product Type | Chemical and Hazardous Materials Sign |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity & Availability | |||
19-035-467
|
Brady™
84436 |
Polyester | 10 in. | 14 in. |
Each for $66.00
|
|
|||
19-035727
|
Brady™
84434 |
Polyester | 3.5 in. | 5 in. |
Each for $13.50
|
|
|||
19-035-726
|
Brady™
22338 |
Plastic | 10 in. | 14 in. |
Each for $36.25
|
|
|||
19-035-725
|
Brady™
22337 |
Plastic | 7 in. | 10 in. |
Each for $31.50
|
|
|||
Description
- Bright colors and bold text provide high visibility to critical messages
- Sign reads “DANGER POISON” with pictogram
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
1 Ea. | |
-40°F | |
DANGER Poison | |
Chemical and Hazardous Materials Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title