Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Chemical and Hazardous Materials Signs: DANGER - FLAMMABLE MATERIALS
Helps identify plant's chemical storage and hazardous materials areas
$22.25 - $98.00
Specifications
Certifications/Compliance | ISO 9001, ANSI Z535, OSHA 1910.145 |
---|---|
For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
Displayed Text | Flammable Material |
Product Type | Hazardous Materials Sign |
Catalog Number | Mfr. No. | Height (Metric) | Material | Width (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (Metric) | Material | Width (Metric) | Price | Quantity & Availability | |||
17-046-22
|
Brady™
72246 |
25.40 cm | Fiberglass | 35.56 cm |
Each for $98.00
|
|
|||
19-017-216
|
Brady™
85184 |
25.40 cm | Polyester | 35.56 cm |
Each for $79.00
|
|
|||
19-035964
|
Brady™
25667 |
25.40 cm | Plastic | 35.56 cm |
Each for $36.75
|
|
|||
19-035965
|
Brady™
85183 |
25.40 cm | Polyester | 25.40 cm |
Each for $22.25
|
|
|||
19-000-659
|
Brady™
43244 |
25.4 cm | Aluminum | 35.5 cm |
Each for $79.00
|
|
|||
Description
- Available in three types of materials: fiberglass, self-sticking polyester and plastic
- Available in various widths and heights
- Black/red on white
Specifications
ISO 9001, ANSI Z535, OSHA 1910.145 | |
Designed for use in industrial, utility, commercial and institutional environments | |
Flammable Material | |
Hazardous Materials Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title